Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300008472.mm.1 |
NM_001079830.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008472.mm.1 |
NM_053170.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008472.mm.1 |
TC0300001049.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008471.mm.1 |
NM_001079830.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008471.mm.1 |
NM_053170.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008471.mm.1 |
TC0300001049.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300008470.mm.1 |
NM_001079830.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008470.mm.1 |
NM_053170.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300008470.mm.1 |
TC0300001049.mm.1 |
MTA TC |