Assay Details
| Gene Symbol: Sike1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025679.3 | NP_079955.1 | 2 - 3 | 335 | 83 | |
| GenBank mRNA | AK014113.1 | - | 2 - 3 | 336 | 83 | |
| AK014247.1 | - | 2 - 3 | 342 | 83 | ||
| AK017452.1 | - | 2 - 3 | 335 | 83 | ||
| AK017694.1 | - | 2 - 3 | 331 | 83 | ||
| AK019264.1 | - | 2 - 3 | 321 | 83 | ||
| AK033553.1 | - | 2 - 3 | 339 | 83 | ||
| AK035590.1 | - | 2 - 3 | 349 | 83 | ||
| BC004090.1 | - | 2 - 3 | 340 | 3593790 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sike1
Genomic Map