Assay Details
Gene Symbol: Ptpn22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008979.2 | NP_033005.1 | 6 - 7 | 602 | 144 | |
NR_104070.1 | 8 - 9 | 627 | 144 | |||
GenBank mRNA | AK153812.1 | - | 8 - 9 | 628 | 144 | |
BC055377.1 | - | 6 - 7 | 524 | 30077916 | 144 | |
M90388.1 | - | 6 - 7 | 587 | 144 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
