Assay Details
Gene Symbol: Gm13125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115077.1 | NP_001108549.1 | 2 - 3 | 863 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2136793 | NM_001115077.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2045662 | NM_001115077.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747506 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18343 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2136793 | NM_001115077.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2045662 | NM_001115077.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23378224 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378238 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03741863 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03430386 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03384313 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03384310 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378232 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03544560 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03408920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03781291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378250 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03544567 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378234 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03384306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03384314 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03377393 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378231 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03781294 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03516757 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03486011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03715549 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378208 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378277 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378212 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03544556 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03544557 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378216 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378229 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03516759 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03486010 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03221857 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378207 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04045769 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23378259 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10518199 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10518200 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10518198 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10518197 | - | - |
Affymetrix Mouse Clariom S Array | PSR0400032642.mm.1 | TC0400003941.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400032642.mm.1 | NM_001115077.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400032642.mm.1 | NM_001115077.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400032643.mm.1 | TC0400003941.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400032643.mm.1 | NM_001115077.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400032643.mm.1 | NM_001115077.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400032644.mm.1 | TC0400003941.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400032644.mm.1 | NM_001115077.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400032644.mm.1 | NM_001115077.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 754775 | - | - |
