Assay Details
Gene Symbol: Brd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033274.3 | NP_001028446.2 | 4 - 5 | 1963 | 61 | |
GenBank mRNA | AK149714.1 | - | 4 - 5 | 1975 | 61 | |
AK220233.1 | - | 3 - 4 | 884 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
