Assay Details
Gene Symbol: Brd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033274.3 | NP_001028446.2 | 9 - 10 | 3297 | 85 | |
GenBank mRNA | AK220233.1 | - | 8 - 9 | 2218 | 85 | |
BC037388.1 | - | 4 - 5 | 624 | 85 | ||
BC049267.1 | - | 2 - 3 | 555 | 85 | ||
BC057182.1 | - | 2 - 3 | 221 | 85 | ||
BC130273.1 | - | 7 - 8 | 1671 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
