Assay Details
| Gene Symbol: Thoc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008425.1 | NP_001008425.1 | 9 - 10 | 856 | 67 | |
| GenBank mRNA | AK089746.1 | - | 6 - 7 | 1184 | 67 | |
| BC085131.1 | - | 9 - 10 | 856 | 5715435 | 67 | |
| BC089551.1 | - | 10 - 11 | 856 | 30303151 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Thoc6
Genomic Map