Assay Details
Gene Symbol: Cldn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018777.4 | NP_061247.1 | 1 - 2 | 40 | 57 | |
GenBank mRNA | AK010560.1 | - | 1 - 2 | 41 | 57 | |
AK010682.1 | - | 1 - 2 | 35 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK010560.1, AK010682.1, NM_018777.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
