Assay Details
Gene Symbol: Cldn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018777.4 | NP_061247.1 | 2 - 3 | 113 | 64 | |
GenBank mRNA | AK010560.1 | - | 2 - 3 | 114 | 64 | |
AK010682.1 | - | 2 - 3 | 108 | 64 | ||
BC005718.1 | - | 2 - 3 | 90 | 3594823 | 64 | |
BC050138.1 | - | 2 - 3 | 96 | 30028814 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK010560.1, AK010682.1, BC005718.1, BC050138.1, NM_018777.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
