Assay Details
Gene Symbol: Zfp213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033496.3 | NP_001028668.2 | 3 - 4 | 897 | 70 | |
GenBank mRNA | AK133902.1 | - | 3 - 4 | 795 | 70 | |
AK147992.1 | - | 3 - 4 | 801 | 70 | ||
AK186649.1 | - | 1 - 2 | 42 | 70 | ||
BC150777.1 | - | 2 - 3 | 640 | 9087688 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
