| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400032686.mm.1 |
TC0400003953.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032686.mm.1 |
NM_001126316.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032686.mm.1 |
NM_001126316.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032688.mm.1 |
TC0400003953.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032688.mm.1 |
NM_001126316.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032688.mm.1 |
NM_001126316.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032689.mm.1 |
TC0400003953.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032689.mm.1 |
NM_001126316.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032689.mm.1 |
NM_001126316.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032687.mm.1 |
TC0400003953.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400032687.mm.1 |
NM_001126316.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400032687.mm.1 |
NM_001126316.1 |
Refseq |