Assay Details
Gene Symbol: Tmsb15a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030106.2 | NP_084382.1 | 1 - 2 | 105 | 97 | |
GenBank mRNA | AK018998.1 | - | 1 - 2 | 107 | 97 | |
AK028086.1 | - | 1 - 2 | 69 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK018998.1, AK028086.1, NM_030106.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
