Assay Details
| Gene Symbol: Capsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029341.1 | NP_083617.2 | 3 - 4 | 459 | 120 | |
| GenBank mRNA | AK006467.1 | - | 3 - 4 | 459 | 120 | |
| BC037621.1 | - | 2 - 3 | 238 | 120 | ||
| BC061015.1 | - | 3 - 4 | 417 | 30297953 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Capsl
Genomic Map