| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500017288.mm.1 |
NM_028003.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017288.mm.1 |
NM_028003.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017288.mm.1 |
TC1500002154.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500017286.mm.1 |
NM_028003.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017286.mm.1 |
NM_028003.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017286.mm.1 |
TC1500002154.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500017287.mm.1 |
NM_028003.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017287.mm.1 |
NM_028003.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017287.mm.1 |
TC1500002154.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500017289.mm.1 |
NM_028003.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017289.mm.1 |
NM_028003.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500017289.mm.1 |
TC1500002154.mm.1 |
MTA TC |