Assay Details
Gene Symbol: Col2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113515.2 | NP_001106987.2 | 10 - 11 | 793 | 66 | |
NM_031163.3 | NP_112440.2 | 11 - 12 | 997 | 66 | ||
GenBank mRNA | AK132311.1 | - | 10 - 11 | 716 | 66 | |
BC051383.1 | - | 10 - 11 | 704 | 6312433 | 66 | |
BC052326.1 | - | 10 - 11 | 683 | 6315379 | 66 | |
BC082331.1 | - | 11 - 12 | 930 | 30537724 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
