Assay Details
| Gene Symbol: Ngfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033217.3 | NP_150086.2 | 3 - 4 | 715 | 147 | |
| GenBank mRNA | AK037248.1 | - | 3 - 4 | 716 | 147 | |
| BC038365.1 | - | 3 - 4 | 706 | 5367638 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ngfr
Genomic Map