Assay Details
| Gene Symbol: Cct6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291242.1 | NP_001278171.1 | 12 - 13 | 1614 | 112 | |
| NM_009839.3 | NP_033969.2 | 12 - 13 | 1545 | 112 | ||
| GenBank mRNA | AK076639.1 | - | 12 - 13 | 1615 | 112 | |
| BC100457.1 | - | 12 - 13 | 1512 | 30918851 | 112 | |
| CF551313.1 | - | 4 - 5 | 431 | 112 | ||
| Z50192.1 | - | 12 - 13 | 1530 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct6b
Genomic Map