Assay Details
| Gene Symbol: Cct6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291242.1 | NP_001278171.1 | 6 - 7 | 892 | 158 | |
| NM_009839.3 | NP_033969.2 | 6 - 7 | 823 | 158 | ||
| GenBank mRNA | AK076639.1 | - | 6 - 7 | 893 | 158 | |
| BC100457.1 | - | 6 - 7 | 790 | 30918851 | 158 | |
| Z50192.1 | - | 6 - 7 | 808 | 158 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct6b
Genomic Map