Assay Details
| Gene Symbol: Cct6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291242.1 | NP_001278171.1 | 9 - 10 | 1227 | 76 | |
| NM_009839.3 | NP_033969.2 | 9 - 10 | 1158 | 76 | ||
| GenBank mRNA | AK076639.1 | - | 9 - 10 | 1228 | 76 | |
| BC100457.1 | - | 9 - 10 | 1125 | 30918851 | 76 | |
| CF551313.1 | - | 1 - 2 | 44 | 76 | ||
| Z50192.1 | - | 9 - 10 | 1143 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct6b
Genomic Map