Assay Details
| Gene Symbol: Unc45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178680.4 | NP_848795.3 | 17 - 18 | 2363 | 59 | |
| GenBank mRNA | AF539793.1 | - | 16 - 17 | 2262 | 59 | |
| AK052294.1 | - | 17 - 18 | 2290 | 59 | ||
| AK081821.1 | - | 17 - 18 | 2302 | 59 | ||
| AK084418.1 | - | 17 - 18 | 2331 | 59 | ||
| AK168968.1 | - | 17 - 18 | 2332 | 59 | ||
| BC084585.1 | - | 17 - 18 | 2296 | 30429260 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Unc45b
Genomic Map