Assay Details
| Gene Symbol: Tnfrsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009401.2 | NP_033427.1 | 1 - 2 | 129 | 96 | |
| GenBank mRNA | AK044269.1 | - | 147 | 96 | ||
| AK044614.1 | - | 147 | 96 | |||
| BC130026.1 | - | 1 - 2 | 120 | 40140833 | 96 | |
| U25416.1 | - | 1 - 2 | 129 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC130026.1, NM_009401.2, U25416.1 transcripts.
Set Membership: |
|

Tnfrsf8
Genomic Map