Assay Details
| Gene Symbol: Tnfrsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009401.2 | NP_033427.1 | 3 - 4 | 328 | 92 | |
| GenBank mRNA | AK044269.1 | - | 346 | 92 | ||
| AK044614.1 | - | 346 | 92 | |||
| BC130026.1 | - | 3 - 4 | 319 | 40140833 | 92 | |
| U25416.1 | - | 3 - 4 | 328 | 92 | ||
More Information
Set Membership: |
|

Tnfrsf8
Genomic Map