Assay Details
| Gene Symbol: Mrpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024227.3 | NP_077189.2 | 4 - 5 | 722 | 74 | |
| GenBank mRNA | AK003727.2 | - | 4 - 5 | 703 | 74 | |
| AK031701.1 | - | 4 - 5 | 692 | 74 | ||
| AK046544.1 | - | 4 - 5 | 703 | 74 | ||
| AK163190.1 | - | 4 - 5 | 705 | 74 | ||
| BC002293.1 | - | 4 - 5 | 74 | |||
| BC005797.1 | - | 4 - 5 | 694 | 74 | ||
| BC080753.1 | - | 4 - 5 | 668 | 74 | ||
| BC100322.1 | - | 4 - 5 | 694 | 74 | ||
| BC145522.1 | - | 4 - 5 | 701 | 74 | ||
| BC145680.1 | - | 4 - 5 | 691 | 40130983 | 74 | |
| BC145682.1 | - | 4 - 5 | 691 | 40130985 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31108448 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31108445 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31108446 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31108444 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P180335 | BC100322.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P180335 | BC145522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P180335 | BC145680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P180335 | BC145682.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P180335 | AK031701.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P180335 | BC100322.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P180335 | BC145522.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P180335 | BC145680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P180335 | BC145682.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P180335 | AK031701.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 240915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878437 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P180335 | BC100322.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P180335 | BC145522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P180335 | BC145680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P180335 | BC145682.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P180335 | AK031701.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P180335 | BC100322.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P180335 | BC145522.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P180335 | BC145680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P180335 | BC145682.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P180335 | AK031701.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180335 | BC100322.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180335 | BC145522.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180335 | BC145680.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180335 | BC145682.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180335 | AK031701.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180335 | BC100322.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180335 | BC145522.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180335 | BC145680.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180335 | BC145682.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P180335 | AK031701.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273197 | BC002293.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31108451 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03296632 | - | - |
| Whole Mouse Genome G4122A | A_51_P273197 | BC002293.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180335 | BC100322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180335 | BC145522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180335 | BC145680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180335 | BC145682.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180335 | AK031701.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180335 | BC100322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180335 | BC145522.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180335 | BC145680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180335 | BC145682.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P180335 | AK031701.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138003 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134238 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13552168 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13552147 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13552173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31108445 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31108446 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31108448 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31108444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31108371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13373412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13552138 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13361599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13361595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13227503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13576325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13576327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13552173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13361593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13227505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13373414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31108449 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10442949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442951 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442950 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442952 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10442953 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416284_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437622_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456313_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10442948 | - | - |
| Mouse Genome 430 2.0 Array | 1416284_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456313_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437622_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1416284_at | - | - |
| Mouse Genome 430 2.0 Array | 1437622_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1456313_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68611_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68611_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700003397.mm.1 | NM_024227.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700003397.mm.1 | NM_024227.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700003397.mm.1 | TC1700000403.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700003398.mm.1 | NM_024227.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700003398.mm.1 | NM_024227.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700003398.mm.1 | TC1700000403.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456313_PM_x_at | - | - |
| Mouse Expression Set 430 | 1437622_x_at | - | - |
| Mouse Expression Set 430 | 1416284_at | - | - |
| Mouse Expression Set 430 | 1416284_PM_at | - | - |
| Mouse Expression Set 430 | 1437622_PM_x_at | - | - |
| Mouse Expression Set 430 | 1456313_x_at | - | - |
| Murine Genome U74v2 Set | 96652_at | - | - |
| Murine Genome U74v2 Set | 96652_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437622_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456313_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416284_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437622_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416284_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456313_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437622_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416284_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456313_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456313_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437622_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416284_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3520673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003520673 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237265 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237265 | - | - |
| mouse-6 v1.1 (Illumina) | scl50963.5.1_38-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3520673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50963.5.1_38-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3520673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1237265 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1237265 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50963.5.1_38-S | - | - |

Mrpl28
Genomic Map