Assay Details
| Gene Symbol: Arhgdig | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008113.3 | NP_032139.1 | 2 - 3 | 467 | 90 | |
| GenBank mRNA | AK144321.1 | - | 2 - 3 | 319 | 90 | |
| BC002032.1 | - | 2 - 3 | 464 | 3490167 | 90 | |
| BC158003.1 | - | 2 - 3 | 332 | 9007312 | 90 | |
| L42463.1 | - | 2 - 3 | 298 | 90 | ||
| U73198.1 | - | 2 - 3 | 307 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgdig
Genomic Map