Assay Details
Gene Symbol: Ergic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026170.3 | NP_080446.1 | 8 - 9 | 737 | 67 | |
GenBank mRNA | AB093571.1 | - | 8 - 9 | 636 | 67 | |
AK003239.1 | - | 8 - 9 | 723 | 67 | ||
AK004629.1 | - | 8 - 9 | 691 | 67 | ||
AK040727.1 | - | 5 - 6 | 889 | 67 | ||
AK084385.1 | - | 8 - 9 | 684 | 67 | ||
AK088157.1 | - | 8 - 9 | 692 | 67 | ||
AK151239.1 | - | 8 - 9 | 691 | 67 | ||
AK173123.1 | - | 8 - 9 | 726 | 67 | ||
BC083144.1 | - | 8 - 9 | 657 | 30432732 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
