Assay Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081069.1 | NP_001074538.1 | 10 - 11 | 704 | 89 | |
| GenBank mRNA | AF061934.1 | - | 9 - 10 | 609 | 89 | |
| AK080707.1 | - | 10 - 11 | 795 | 89 | ||
More Information
Set Membership: |
|

Rgs11
Genomic Map