Assay Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081069.1 | NP_001074538.1 | 11 - 12 | 758 | 63 | |
| GenBank mRNA | AF061934.1 | - | 10 - 11 | 663 | 63 | |
| AK039314.1 | - | 10 - 11 | 1380 | 63 | ||
| AK080707.1 | - | 11 - 12 | 849 | 63 | ||
| AK161551.1 | - | 7 - 8 | 817 | 63 | ||
| BC049910.1 | - | 9 - 10 | 1462 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs11
Genomic Map