Assay Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081069.1 | NP_001074538.1 | 2 - 3 | 176 | 71 | |
| GenBank mRNA | AF061934.1 | - | 1 - 2 | 81 | 71 | |
| AK039314.1 | - | 2 - 3 | 145 | 71 | ||
| BC037655.1 | - | 2 - 3 | 145 | 71 | ||
| BC049910.1 | - | 2 - 3 | 162 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rgs11
Genomic Map