Assay Details
Gene Symbol: Ksr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013571.2 | NP_038599.1 | 16 - 17 | 2351 | 62 | |
GenBank mRNA | AK137532.1 | - | 15 - 16 | 2088 | 62 | |
U43585.1 | - | 16 - 17 | 2358 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
