Assay Details
Gene Symbol: Ksr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013571.2 | NP_038599.1 | 2 - 3 | 434 | 72 | |
GenBank mRNA | AK036566.1 | - | 2 - 3 | 490 | 72 | |
AK164833.1 | - | 2 - 3 | 554 | 71 | ||
U43585.1 | - | 2 - 3 | 441 | 72 | ||
X81634.1 | - | 2 - 3 | 216 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
