Assay Details
| Gene Symbol: Lgals9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010708.2 | NP_034838.2 | 5 - 6 | 648 | 102 | |
| GenBank mRNA | U55061.1 | - | 5 - 6 | 555 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lgals9
Genomic Map