Assay Details
| Gene Symbol: Oxct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024188.6 | NP_077150.1 | 3 - 4 | 622 | 107 | |
| GenBank mRNA | AB085609.1 | - | 3 - 4 | 364 | 107 | |
| AK009027.1 | - | 3 - 4 | 402 | 107 | ||
| AK010790.1 | - | 3 - 4 | 365 | 107 | ||
| AK011354.1 | - | 3 - 4 | 302 | 107 | ||
| AK078086.1 | - | 3 - 4 | 106 | |||
| AK146158.1 | - | 3 - 4 | 380 | 107 | ||
| AK146342.1 | - | 3 - 4 | 367 | 107 | ||
| AK151662.1 | - | 3 - 4 | 361 | 107 | ||
| AK151695.1 | - | 3 - 4 | 352 | 107 | ||
| AK167681.1 | - | 3 - 4 | 452 | 107 | ||
| BC003422.1 | - | 3 - 4 | 326 | 2598824 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oxct1
Genomic Map