Assay Details
| Gene Symbol: Rps6kb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114334.1 | NP_001107806.1 | 7 - 8 | 794 | 92 | |
| GenBank mRNA | AK012045.1 | - | 2 - 3 | 173 | 92 | |
| AK032724.1 | - | 7 - 8 | 756 | 92 | ||
| AK135716.1 | - | 7 - 8 | 762 | 92 | ||
| AK163258.1 | - | 7 - 8 | 764 | 92 | ||
| BC038491.1 | - | 7 - 8 | 767 | 4010710 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6kb1
Genomic Map