Assay Details
Gene Symbol: Dnajb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021317.2 | NP_067292.2 | 1 | 934 | 109 | |
GenBank mRNA | AK006201.1 | - | 1 | 960 | 109 | |
AK077186.1 | - | 1 | 963 | 109 | ||
BC048738.1 | - | 1 | 923 | 109 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80423 | BC048738.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80423 | BC104396.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80423 | BC104397.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80423 | BC119478.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80423 | AK077186.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80423 | BC048738.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80423 | BC104396.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80423 | BC104397.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80423 | BC119478.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P80423 | AK077186.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153453 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80423 | BC048738.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80423 | BC104396.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80423 | BC104397.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80423 | BC119478.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80423 | AK077186.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80423 | BC048738.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80423 | BC104396.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80423 | BC104397.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80423 | BC119478.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P80423 | AK077186.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377814 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377812 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377802 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377785 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377786 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377809 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377792 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377781 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377789 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377780 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377807 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377816 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377818 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377800 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377793 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377795 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377815 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377805 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377810 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377783 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30377798 | - | - |
Whole Mouse Genome G4122A | A_52_P80423 | BC048738.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80423 | BC104396.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80423 | BC104397.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80423 | BC119478.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80423 | AK077186.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80423 | BC048738.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80423 | BC104396.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80423 | BC104397.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80423 | BC119478.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P80423 | AK077186.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129851 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12100784 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11903014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30377781 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377781 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377783 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377798 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377814 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377823 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377795 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377830 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377817 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377789 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11864281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12182330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377819 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11842874 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377793 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12458914 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377792 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377807 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377821 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377816 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377809 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377786 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12334812 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377815 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377805 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377828 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30377778 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11864284 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10430741 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10430740 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10430742 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450991_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10430739 | - | - |
Mouse Genome 430 2.0 Array | 1450991_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450991_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57755_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57755_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500015100.mm.1 | NM_021317.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015100.mm.1 | NM_021317.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015100.mm.1 | TC1500001921.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500015099.mm.1 | NM_021317.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015099.mm.1 | NM_021317.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015099.mm.1 | TC1500001921.mm.1 | MTA TC |
Mouse Expression Set 430 | 1450991_PM_at | - | - |
Mouse Expression Set 430 | 1450991_at | - | - |
Mouse Genome 430A 2.0 Array | 1450991_at | - | - |
Mouse Genome 430A 2.0 Array | 1450991_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450991_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450991_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1090593 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001090593 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2777208 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2777208 | - | - |
mouse-6 v1.1 (Illumina) | scl46935.1.32_89-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1090593 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl46935.1.32_89-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1090593 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2777208 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2777208 | - | - |
