Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500015004.mm.1 |
NM_001082536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015004.mm.1 |
NM_001082536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015004.mm.1 |
NM_153049.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015004.mm.1 |
TC1500001912.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500015003.mm.1 |
NM_001082536.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015003.mm.1 |
NM_001082536.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015003.mm.1 |
NM_153049.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500015003.mm.1 |
TC1500001912.mm.1 |
MTA TC |