Assay Details
Gene Symbol: Kcnh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010601.3 | NP_034731.3 | 3 - 4 | 448 | 58 | |
GenBank mRNA | AY380579.1 | - | 3 - 4 | 448 | 58 | |
BC141013.1 | - | 3 - 4 | 563 | 9055625 | 58 | |
BC145145.1 | - | 3 - 4 | 563 | 9053696 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
