Assay Details
| Gene Symbol: Sgms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028943.5 | NP_083219.2 | 3 - 4 | 1139 | 89 | |
| GenBank mRNA | AK016659.1 | - | 3 - 4 | 1153 | 89 | |
| BC117782.2 | - | 3 - 4 | 1143 | 40092081 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgms2
Genomic Map