Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400017192.mm.1 |
NM_007549.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017192.mm.1 |
NM_007549.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017192.mm.1 |
TC1400002219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017193.mm.1 |
NM_007549.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017193.mm.1 |
NM_007549.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017193.mm.1 |
TC1400002219.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017191.mm.1 |
NM_007549.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017191.mm.1 |
NM_007549.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017191.mm.1 |
TC1400002219.mm.1 |
MTA TC |