Assay Details
| Gene Symbol: Blk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007549.2 | NP_031575.2 | 7 - 8 | 1017 | 120 | |
| GenBank mRNA | AK089888.1 | - | 7 - 8 | 1017 | 120 | |
| AK171640.1 | - | 7 - 8 | 964 | 120 | ||
| BC030668.1 | - | 7 - 8 | 867 | 1178763 | 120 | |
| CT010240.1 | - | 6 - 7 | 597 | 120 | ||
| M30903.1 | - | 7 - 8 | 946 | 120 | ||
More Information
Set Membership: |
|

Blk
Genomic Map