Assay Details
Gene Symbol: Rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009029.2 | NP_033055.2 | 25 - 26 | 2787 | 93 | |
GenBank mRNA | AK132875.1 | - | 14 - 15 | 1533 | 93 | |
AK141028.1 | - | 25 - 26 | 2819 | 93 | ||
AK146467.1 | - | 20 - 21 | 2118 | 93 | ||
AK148403.1 | - | 1 - 2 | 103 | 93 | ||
BC096525.1 | - | 25 - 26 | 2787 | 6832808 | 93 | |
DQ400415.1 | - | 25 - 26 | 2637 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
