Assay Details
Gene Symbol: Lrrc63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027581.1 | NP_081857.1 | 8 - 9 | 1602 | 112 | |
GenBank mRNA | AK014850.1 | - | 7 - 8 | 1334 | 112 | |
BC145796.1 | - | 8 - 9 | 1597 | 40131099 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
