Assay Details
| Gene Symbol: Cd46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010778.3 | NP_034908.1 | 4 - 5 | 525 | 137 | |
| GenBank mRNA | AB001566.1 | - | 4 - 5 | 502 | 137 | |
| AB010919.1 | - | 4 - 5 | 522 | 137 | ||
| AB022600.1 | - | 4 - 5 | 502 | 137 | ||
| AK006632.1 | - | 4 - 5 | 526 | 137 | ||
| BC150660.1 | - | 4 - 5 | 526 | 9087571 | 137 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd46
Genomic Map