Assay Details
| Gene Symbol: Cd46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010778.3 | NP_034908.1 | 7 - 8 | 965 | 151 | |
| GenBank mRNA | AB001566.1 | - | 7 - 8 | 942 | 151 | |
| AB010919.1 | - | 7 - 8 | 962 | 151 | ||
| AB022600.1 | - | 8 - 9 | 1015 | 151 | ||
| AK006632.1 | - | 7 - 8 | 966 | 151 | ||
| BC150660.1 | - | 7 - 8 | 966 | 9087571 | 151 | |
More Information
Set Membership: |
|

Cd46
Genomic Map