Assay Details
| Gene Symbol: Cr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013499.2 | NP_038527.2 | 5 - 6 | 1146 | 81 | |
| GenBank mRNA | AI098173.1 | - | 5 - 6 | 81 | ||
| AK004825.1 | - | 5 - 6 | 1138 | 81 | ||
| AK134226.1 | - | 920 | 81 | |||
| AK160440.1 | - | 5 - 6 | 821 | 81 | ||
| BC028945.1 | - | 5 - 6 | 913 | 5371665 | 81 | |
| BC092048.1 | - | 5 - 6 | 904 | 3660242 | 81 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cr1l
Genomic Map