Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200009750.mm.1 |
NM_029107.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009750.mm.1 |
NM_029107.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009750.mm.1 |
TC1200001362.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009749.mm.1 |
NM_029107.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009749.mm.1 |
NM_029107.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009749.mm.1 |
TC1200001362.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009748.mm.1 |
NM_029107.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009748.mm.1 |
NM_029107.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009748.mm.1 |
TC1200001362.mm.1 |
MTA TC |