Assay Details
| Gene Symbol: Mfsd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033488.2 | NP_001028660.2 | 7 - 8 | 764 | 88 | |
| GenBank mRNA | AK133061.1 | - | 7 - 8 | 779 | 88 | |
| AK156138.1 | - | 8 - 9 | 916 | 88 | ||
| AK157892.1 | - | 6 - 7 | 1591 | 88 | ||
| AK162620.1 | - | 7 - 8 | 763 | 88 | ||
| AK171660.1 | - | 4 - 5 | 427 | 88 | ||
| AK172423.1 | - | 7 - 8 | 765 | 88 | ||
| BC145584.1 | - | 7 - 8 | 777 | 88 | ||
More Information
Set Membership: |
|

Mfsd2b
Genomic Map