Assay Details
| Gene Symbol: Mfsd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033488.2 | NP_001028660.2 | 8 - 9 | 889 | 77 | |
| GenBank mRNA | AK133061.1 | - | 8 - 9 | 904 | 77 | |
| AK156138.1 | - | 9 - 10 | 1041 | 77 | ||
| AK157892.1 | - | 7 - 8 | 1716 | 77 | ||
| AK162620.1 | - | 8 - 9 | 888 | 77 | ||
| AK171660.1 | - | 5 - 6 | 552 | 77 | ||
| AK172423.1 | - | 8 - 9 | 890 | 77 | ||
| BC145584.1 | - | 8 - 9 | 902 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd2b
Genomic Map