Assay Details
| Gene Symbol: Gpr137c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027518.2 | NP_081794.2 | 5 - 6 | 1251 | 63 | |
| GenBank mRNA | AY255546.1 | - | 1 - 2 | 75 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr137c
Genomic Map