Assay Details
| Gene Symbol: Ctsc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009982.4 | NP_034112.3 | 1 - 2 | 281 | 89 | |
| GenBank mRNA | AK002454.1 | - | 1 - 2 | 240 | 89 | |
| AK049406.1 | - | 1 - 2 | 240 | 89 | ||
| AK146858.1 | - | 1 - 2 | 289 | 89 | ||
| AK150761.1 | - | 1 - 2 | 241 | 89 | ||
| AK151388.1 | - | 1 - 2 | 240 | 89 | ||
| AK151711.1 | - | 1 - 2 | 241 | 89 | ||
| AK151861.1 | - | 1 - 2 | 127 | 89 | ||
| AK152524.1 | - | 1 - 2 | 238 | 89 | ||
| AK153168.1 | - | 1 - 2 | 241 | 89 | ||
| AK167882.1 | - | 1 - 2 | 240 | 89 | ||
| BC067063.1 | - | 1 - 2 | 214 | 30541061 | 89 | |
| U74683.3 | - | 1 - 2 | 238 | 89 | ||
| U89269.1 | - | 1 - 2 | 239 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctsc
Genomic Map