Assay Details
| Gene Symbol: Akt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011785.3 | NP_035915.3 | 3 - 4 | 337 | 131 | |
| GenBank mRNA | AF124142.1 | - | 3 - 4 | 327 | 131 | |
| BC066861.1 | - | 3 - 4 | 337 | 30089997 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akt3
Genomic Map